missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Proteinase K for MagMAX™-96 DNA Multi-Sample Kit

This is the Proteinase K found in the MagMAX™-96 DNA Multi-Sample Kit, made available separately for applications that require more proteinase K enzyme than is provided in the kit.

Brand:  Invitrogen™ 4489110

61.20 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 15332506

  • £72.00 / 96 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 1 tube
This item is not returnable. View return policy

Description

Description

This is the Proteinase K found in the MagMAX™-96 DNA Multi-Sample Kit (Cat. No. 4413020, 4413021, 4413022), made available separately for applications that require more proteinase K enzyme than is provided in the kit.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 tube

Store at -25°C to -15°C.
7500 Fast System, 7500 System, 7900HT Fast System, 7900HT System, Ion PGM™ System, KingFisher™ Flex Magnetic Particle Processor, OpenArray™, MagMAX™ Express-96 Standard Magnetic Particle Processor, MagMAX™ Express-96 Deep Well Magnetic Particle Processor
High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
DNA Multi-Sample Kit
DNase-Free
Liquid
Genomic DNA
Next-Generation Sequencing, Real-Time Quantitative PCR (qPCR)
MagMAX™
96 preps
Genomic DNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.