missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Recombinant Proteinase K Solution (20 mg/mL)

Product Code. p-4926917
Click to view available options
Quantity:
1.25 mL
5 x 1.25 mL
Unit Size:
6.25mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10665795

Brand: Invitrogen™ AM2546

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Recombinant Proteinase K is a nonspecific serine protease that is useful for general digestion of proteins.

Recombinant Proteinase K is a nonspecific serine protease that is useful for general digestion of proteins.

Proteinase K remains active:

  • Over a wide pH range (optimal activity between 6.5 and 9.5)
  • Under denaturing conditions (e.g., in the presence of SDS or urea)
  • In the presence of metal chelating agents (e.g., EDTA)
  • At comparatively high temperatures (optimum digestion temperature is 65°C)

Applications

Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and quality testing

Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit definition

One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within one minute at 37°C using hemoglobin as a substrate.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 mg/mL
Host Species T. album
Content And Storage Store at -20°C.
Shipping Condition Dry Ice
Quantity 1.25 mL
Product Type Proteinase K
Product Line Ambion
Form Liquid
compliance-icons
Product Identifier
  • PROTEINASE K (20MG/ML) 1.25ML
Signal Word
  • Danger
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Proteinase, tritirachiumalbum serine

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.