Learn More
Invitrogen™ PureLink™ DNase Set
The RNase-free DNase set comes lyophilized and is optimized for on-column digestion of DNA using PureLink™ protocols.
Brand: Invitrogen™ 12185010
67.33 GBP valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Includes: PureLink™ DNase (1500U, lyophilized), 10X DNase I Reaction Buffer (1mL), RNase-Free Water (1mL)
Description
Using the PureLink™ DNase Set
The RNase-free DNase set comes lyophilized and is optimized for on-column digestion of DNA using PureLink™ protocols. The degree of DNA digestion performed by PureLink™ DNase is optional for most PureLink™ RNA kits, due to the efficient removal of the majority of DNA by the PureLink™ technology; however, some downstream applications that are sensitive to small amounts of DNA may require further removal of DNA.
- RNase-free for use in RNA purification
- Optimized for on-column digestion
- Lyophilized for stability
Order Info
Shipping Condition: Room Temperature

Specifications
10X | |
50 Preps | |
Room Temperature |
Contains: • PureLink™ DNase (1500 U, lyophilized) • 10X DNase I Reaction Buffer (1 mL) • RNase-Free Water (1 mL) Store at 4°C upon receipt. After reconstituted, PureLink™ DNase should be stored at -20°C. |
|
50 Preps |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
3116038 | Certificate of Analysis | 31/01/2025 | 12185010 |
3048827 | Certificate of Analysis | 31/01/2025 | 12185010 |
3088764 | Certificate of Analysis | 31/01/2025 | 12185010 |
2485862 | Certificate of Analysis | 29/01/2025 | 12185010 |
2535427 | Certificate of Analysis | 29/01/2025 | 12185010 |
Special Offers
-
Multiple Manufacturers
More Discovery, Less Cost
Save on thousands of eligible products.Promo Code: 24111Expires: 2025-03-21
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.