Learn More
Invitrogen™ Ambion™ Recombinant RNase A
Rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity
Brand: Invitrogen™ AM2269
134.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
For large batch sizes, use of RNase is still only effective technique applicable to remove host RNA. Other methods such as selective precipitation, gel filtration, chromatography, and endogenous E. coli RNase digestion have only been effective for small-scale or bench-scale purification. This product was designed specifically to minimize the risk of introducing pathogens into bioprocessing.
- Substantially free of host RNA
- For research applications
DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Plasmid DNA Purification
Order Info
Shipping Conditions: Dry ice
Specifications
-20°C | |
RNase | |
Recombinant RNase A |
Dry Ice | |
200 μg | |
Ambion™ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.