missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNAlater™-ICE Frozen Tissue Transition Solution

Product Code. p-4918685
Click to view available options
Quantity:
25 mL
10 x 25 mL
Unit Size:
250mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10584445

Brand: Invitrogen™ AM7030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

RNAlater-ICE is a novel reagent for transitioning frozen tissue to a state that is easily processed for extraction of high quality RNA.

RNAlater-ICE is a novel reagent for transitioning frozen tissue to a state that is easily processed for extraction of high quality RNA. Frozen tissues are simply submerged in RNAlater-ICE and allowed to thaw overnight at -20°C. Once thawed, the tissues can be processed like fresh tissues using standard RNA isolation procedures. No more laborious grinding of frozen tissue to safeguard the RNA.

• Process previously frozen tissues like freshly harvested samples
• Thawing tissues in RNAlater-ICE protects RNA from degradation
• No more tissue pulverizing with mortar and pestle and awkward transfer of powder to tube
• Easily apportion frozen tissue samples for multiple experiments
• Quick Freezing Tissues Preserves RNA

Often, tissues that need to be stored prior to RNA isolation are “snap” or “flash” frozen on dry ice or in liquid nitrogen to preserve RNA integrity. RNA in tissue is stable while frozen at -80°C but thawing the tissue prior to or during its disruption can result in RNA degradation. This is true even if the tissue thaws while in the denaturation solution.

Processing Frozen Tissues is Problematic
Frozen tissues are typically ground with a chilled mortar and pestle in order to maintain RNA quality. Liquid nitrogen must be added to the mortar to keep the sample frozen while it is ground. For multiple samples this process is laborious. Either a separate mortar and pestle set is needed for each sample, or the set must be thawed and cleaned after each sample is processed to avoid cross-contamination. Powdered tissue can also thaw during transfer to a homogenization vessel. This often results in formation of clumps that do not readily disperse in the lysis solution, resulting in RNA degradation and loss. Very small samples should be homogenized immediately in lysis solution, which can again be cumbersome if there are multiple samples to process.

Process Frozen Tissue Without Jeopardizing RNA Integrity
RNAlater-ICE solves all of these problems. Simply submerge frozen tissue samples in 10 volumes of RNAlater-ICE and store overnight at -20°C (the solution will remain liquid at these temperatures). As the tissue thaws, RNA integrity is protected. Once treated, tissue can be safely stored at 4°C or even at room temperature (for a limited period of time) and can be further dissected or processed prior to homogenization in a standard RNA isolation lysis buffer. Thus the same frozen tissue sample can be used multiple times for different experiments without compromising RNA integrity.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 25 mL RNAlater-ICE Solution; room temperature

Description RNA Storage Reagent
Sample Type Tissue (Frozen)
Quantity 25 mL
Shipping Condition Room Temperature
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Frozen Tissue Transition Solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, methanol, isopropanol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.