Learn More
Invitrogen™ RNAqueous™-Micro Total RNA Isolation Kit
The RNAqueous™-Micro Kit is used for phenol-free total RNA isolation from micro-sized samples (e.g., from less than 500,000 cultured cells, from few as 10 laser capture microdissected cells, or from ≤10 mg of tissue) using a guanidinium-based lysis/denaturant and glass fiber filter separation technology.
Brand: Invitrogen™ AM1931
270.00 GBP valid until 2024-09-27
Use promo code "21650" to get your promotional price.
Includes: 50 Micro Filter Cartridge Assembly; 50 Micro Elution Tubes; 15mL Wash Solution 1 Concentrate; 28mL Wash Solution 2/3 concentrate; 25 Lysis Solution; 200μL LCM Additive; 55μL DNase 2U/μL; 600μL 10X DNase I Buffer; 150μL DNase Inactivation Reagent; 5mL Elution Solution
Description
The RNAqueous™-Micro Kit is used for phenol-free total RNA isolation from micro-sized samples (e.g., from less than 500,000 cultured cells, from few as 10 laser capture microdissected cells, or from ≤10 mg of tissue) using a guanidinium-based lysis/denaturant and glass fiber filter separation technology. DNA-free™ DNA removal reagents are included in the kit, making the isolated RNA especially suitable for RT-PCR. This kit provides:
• Highly concentrated RNA—for increased sensitivity in downstream assays
• Isolation of high-quality RNA from micro-scale samples
• Reagents/protocol for RNA recovery from LCM samples
How it works
Samples are solubilized in a powerful chaotropic solution, and RNA is purified on a glass fiber filter cartridge specifically designed for very small samples. A simple modification of the protocol allows recovery of both large and small RNA species including tRNAs, 5S rRNA, and microRNAs. The RNA is eluted in a minimal volume of 20 μL. Residual genomic DNA is efficiently removed using the DNA-free™ system included in the kit. The RNAqueous™-Micro Kit contains enough reagents for 50 RNA isolations of DNA-free RNA.
Not high-throughput compatible (Manual)
Applications
DNA and RNA Purification and Analysis, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
• 50 Filter Cartridges with Collection Tubes; store at room temperature • 50 Elution Tubes (1.5 mL); room temperature • 15 mL Wash Solution 1 (concentrated); room temperature • 28 mL Wash Solution 2/3 (concentrated); 4°C • 25 mL Lysis Solution; 4°C • 200 μL LCM Additive; -20°C • 55 μL DNase; -20°C • 600 μL 10X DNase I Buffer; -20°C • 150 μL DNase Inactivation Reagent; -20°C • 5 mL Elution Solution; room temperature |
|
15 min. (After sample homogenization) | |
Cells, Tissue, Laser Capture Microdissection (LCM) | |
Not High-throughput Compatible (Manual) | |
50 Preps | |
Cells: ≤105 Tissue: ≤10 mg |
Silica Spin Column | |
10–20 μL | |
Total RNA | |
RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling | |
Box 1: Dry Ice Box 2: Room Temperature |
Safety and Handling
- RNAqueous - microkit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H332-Harmful if inhaled.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 2-mercaptoethanol, thiocyanic acid
For Research Use Only. Not for use in diagnostic procedures.