Learn More
Invitrogen™ Ambion™ RNase A, affinity purified, 1 mg/mL
Rigorously tested for contaminating nonspecific endonuclease, exonuclease, and protease activity
£114.00 - £334.00
Specifications
Concentration | 1 mg/mL |
---|---|
Content And Storage | -20°C |
Shipping Condition | Dry Ice |
Enzyme | RNase |
Product Type | RNase A |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10095064
|
Invitrogen™
AM2270 |
200 μg |
£114.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10544255
|
Invitrogen™
AM2271 |
1 mg |
£334.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Ambion RNase A is endonuclease that specifically cleaves 3' of U and C residues. Affinity Purified RNase A is intended for critical applications when absence of DNase and other nonspecific nuclease activities is essential.
- Supplied in one tube containing 200μg (1mg/mL)
- Preparation contains RNase B, a carbohydrate isoform of RNase A
DNA & RNA Purification & Analysis, DNA Extraction, Maxiprep, Midiprep, Miniprep, Nuclease Protection Assays, Nucleic Acid Gel Electrophoresis & Blotting, Plasmid DNA Purification
Order Info
Shipping Conditions: Dry ice
Specifications
1 mg/mL | |
Dry Ice | |
RNase A |
-20°C | |
RNase | |
Ambion™ |
For Research Use Only. Not for use in diagnostic procedures.