missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RNase AWAY™ Decontamination Reagent
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
Description
- Ready-to-use solution for eliminating RNase and DNA contamination from labware
- Apply evenly over surface of glassware or plasticware to be treated and then rinse it away with distilled water
- Unwanted RNase and DNA contamination are eliminated
- Nonabrasive, noncarcinogenic, and nonbiologically corrosive
Order Info
Shipping Condition: Room temperature
Specifications
Specifications
| Format | Liquid |
| Content And Storage | Store at room temperature. |
| Shipping Condition | Room Temperature |
| Quantity | 250 mL |
| Reagent Type | RNase Control Reagent |
| Product Line | RNase Away |
| For Use With (Application) | RNA Extraction |
Safety and Handling
Product Identifier
- RNASE AWAY
- Warning
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P362-Take off contaminated clothing.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction