missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ RNase Inhibitor, cloned, 40 U/μL

Binds neutral pancreatic RNase A type enzymes

£184.00 - £496.00

Specifications

Concentration 100 mM
Content And Storage Store at -20°C. Do not store in a frost–free freezer.
Product Line Ambion™
Shipping Condition Dry Ice
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10177094
View Documents
Invitrogen™
AM2682
2500 U
£184.00
Each
Please to purchase this item. Need a web account? Register with us today!
10430725
View Documents
Invitrogen™
AM2684
10,000 U
£496.00
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ambion RNase Inhibitor, a recombinant human protein produced in E. coli, is a potent inhibitor of neutral pancreatic RNase A type enzymes. The mode of inhibition is noncompetitive; the inhibitor tightly binds RNases in a 1:1 ratio. The enzyme has been shown to inactivate RNases present in many tissues and cell types. Addition of the ribonuclease inhibitor (RI) has been shown to be useful whenever the integrity of RNA must be maintained such as in the preparation of cDNA by reverse transcription, in vitro RNA transcription, and in in vitro protein synthesis. RI does not inhibit RNase I, T1, T2, H, U1, U2, or CL3. RI requires a minimum of 1mM DTT to maintain activity, and has an activity range between pH 5.0 to 8.0 with maximal activity between pH 7.0 and 8.0. Since the mode of inhibition is the formation of a 1:1 complex with RNases, care must be taken to avoid denaturation or oxidation of the ribonuclease inhibitor (e.g., by addition of SDS, urea, etc.). RI should be added to transcription, translation, and cDNA synthesis reactions to give a final concentration of 1U/μL. RNase Inhibitor is rigorously tested for contaminating RNase, exonuclease, endonuclease, and protease activity.
Unit Definition: One unit is the amount of protein required to inhibit the activity of 5ng of RNase A by 50%. Unit assay conditions: 100mM Tris-acetate (pH 6.5), 1mM EDTA, 1mM cyclic 2 ft.,3 ft.-CMP, and RNase inhibitor. Addition of RNase A initiates the reaction. RNase Inhibitor activity is measured by the inhibition of hydrolysis of cyclic 2 ft., 3 ft.-CMP by RNase A.

Applications:

Gene Expression Analysis and Genotyping, In Vitro Transcription, PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Condition: Dry ice

Specifications

Specifications

100 mM
Store at -20°C. Do not store in a frost–free freezer.
Ambion™
Dry Ice
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Ambion™ RNase Inhibitor, cloned, 40 U/μL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.