missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RNase T1 (1 U/μL)

Product Code. 10406734
Click to view available options
Quantity:
200 μL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10406734

Brand: Invitrogen™ AM2283

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

RNase T1, 10X RNA Structure Buffer, 1X Alkaline Hydrolysis Buffer, Yeast RNA, Precipitation/Inactivation Buffer, and Gel Loading Buffer II

Invitrogen™ RNase T1, RNA-Grade cleaves 3' of single-stranded G residues and is tested for purity to ensure that no contaminating ribonuclease activities are present that could cleave at unanticipated sites.

Invitrogen™ RNase T1, RNA-Grade cleaves 3′ of single-stranded G residues and is tested for purity to ensure that no contaminating ribonuclease activities are present that could cleave at unanticipated sites. Supplied in one tube of 200μL (1U/μL). RNase T1 can be used to perform boundary experiments to define the minimal RNA sequence required for selectable activities such as protein binding or catalysis. In addition to applications for RNA structural analysis, RNA-Grade ribonucleases can be used to map protein binding sites on RNAs by comparing cleavage patterns in the presence and absence of an RNA binding protein.

  • 100 Units is the amount of enzyme that yields an increase in absorption at 260 nm of 0.01428 units/min. at room temperature using 60 μg/mL yeast total RNA as a substrate.
  • RNase A (Cat. No. AM2274) cleaves 3′ of single-stranded C and U residues. Combinations of the single- and double-stranded specific ribonucleases can provide rapid analysis of the physical structure of an interesting RNA.

Order Info

Shipping Condition: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 U/μL
Content And Storage RNase T1, 10X RNA Structure Buffer, 1X Alkaline Hydrolysis Buffer, Yeast RNA, Precipitation/Inactivation Buffer, and Gel Loading Buffer II

Store all components at –20°C.
Format Tube
Enzyme RNase
Fidelity (vs. Taq) 10X
For Use With (Application) Real Time PCR (qPCR)
No. of Reactions 200
Product Line Ambion
Product Type Rnase T1 PCR Kit
Quantity 200 μL
Shipping Condition Dry Ice
Source RNA
Form Solution
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.