missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ RT-PCR Grade Water

Product Code. 10289774
Click to view available options
Quantity:
10 x 1.5 mL
Unit Size:
17mL
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10289774

Brand: Invitrogen™ AM9935

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

RT-PCR Grade Water is certified free of nucleases, and free of nucleic acid contamination that may cause false-positive signals in RT-PCR. The RT-PCR Grade Water is ready to use and requires no preparation, mixing, or autoclaving.

RT-PCR Grade Water is certified free of nucleases, and free of nucleic acid contamination that may cause false-positive signals in RT-PCR. The RT-PCR Grade Water is ready to use and requires no preparation, mixing, or autoclaving.

Quality control
RT-PCR Grade Water is tested for prokaryotic as well as eukaryotic genomic DNA contamination using an ultrasensitive PCR assay. It is provided in 10 x 1.5 mL tubes. Prokaryotic DNA contamination is assessed by performing PCR with primers to 16S rRNA. Eukaryotic DNA contamination is assessed by performing PCR with primers to 18S rRNA. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity and is suitable for any PCR application.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Grade Molecular Biology
Packaging Tube
Treatment(s) Not DEPC-Treated
Quantity 10 x 1.5 mL
Purification Method Autoclaved, Membrane-Filtered
Shipping Condition Room Temperature
Product Line Ambion

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.