missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Salmon Sperm DNA, sheared (10 mg/mL)

Product Code. 10249194
Click to view available options
Quantity:
10 x 10 mg tubes
Unit Size:
Pack of 10
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10249194

Brand: Invitrogen™ AM9680

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Sheared Salmon Sperm DNA is a very effective blocking agent when used in Northern prehybridization and hybridization buffers at a concentration of 100 to 200 μg/mL.

Sheared Salmon Sperm DNA is a very effective blocking agent when used in Northern prehybridization and hybridization buffers at a concentration of 100 to 200 μg/mL.

Using this product
This sheared salmon sperm DNA has been treated with Proteinase K to remove any contaminating nucleases, followed by organic extraction with phenol:chloroform and ethanol precipitation. Salmon Sperm DNA is rigorously tested for RNase and DNase contamination and is suspended in nuclease-free water in a 1mL tube at a concentration of 10 mg/mL (1 mL per tube).

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store pure solution at –20°C
Concentration 10 mg/mL
For Use With (Application) Block Non-Specific Hybridization
Product Type Salmon Sperm DNA
Product Line Ambion
Quantity 10 x 10 mg tubes
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.