Learn More
Invitrogen™ Ambion™ SP6 RNA Polymerase, cloned, 20 U/μL
A cloned high-purity RNA polymerase with a high specificity for its promoter
Brand: Invitrogen™ AM2071
171.95 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.01000kg
Includes: One tube containing 1000 units at a concentration of 20U/μL
Description
- This enzyme will transcribe large amounts of RNA from DNA sequences downstream of the promoter and show no cross talk between promoters
- One tube containing 1000U at a concentration of 20U/μL is provided. Its high purity makes it ideal for synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA
- The RNA Polymerase is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity
- Functionality of enzyme is tested in In Vitro Transcription reaction under optimal and limiting UTP concentration.
Unit Definition: One unit is the amount of SP6 RNA Polymerase required to catalyze the incorporation of 1 nmol of nucleoside triphosphate in 60 minutes at 37°C
DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, in vitro Transcription, Nucleic Acid Labeling & Oligo Synthesis, RNA Labeling
Order Info
Shipping Condition: Dry ice
Specifications
SP6 RNA Polymerase | |
20 μL | |
20 U/μL | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.