Learn More
Invitrogen™ Spin Columns and Tubes
For rapid elution of DNA from agarose gel slices
Brand: Invitrogen™ AM10065
189.00 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes: 30 columns
Description
Simply excise a band from the gel, place it in the spin column, and centrifuge for five minutes. DNA is forced through the column into the tube below, while agarose is left behind. DNA is ready for subsequent manipulations such as restriction enzyme digestion or ligation.
- No electroelution is needed; columns only require microcentrifuge
- Certified DNase- and RNase-free
Order Info
Shipping Condition: Room Temperature
Specifications
Filtered | |
Tube, Spin Column | |
100 units | |
Store tubes at room temperature. | |
Single-use | |
Invitrogen |
DNA Extraction from Agarose Gels | |
Universal Fit | |
Spin Columns and Tubes | |
Room Temperature | |
Microcentrifuge |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.