missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Stabilized Blood-to-CT™ Nucleic Acid Preparation Kit for qPCR, compatible with Tempus™ Blood RNA Tubes

The Stabilized Blood-to-CT Nucleic Acid Preparation Kit for qPCR makes it possible to perform expression analysis directly from a 500 μL aliquot of human whole blood collected in Tempus Blood RNA tubes without conventional RNA purification.

Brand:  Invitrogen™ 4449080

Code : NEW

Additional Details : Weight : 0.00450kg

Product Code. 11310922

  • £1720.00 / 200 reactions
Estimated Shipment: 05-08-2024
to see stock.

Includes:

Box 1 -

  • Digestion Solution 20mL
  • Tempus Pellet Enhancer 4mL
  • Tempus Wash #1 150mL
  • Tempus Wash #2 150mL
  • 2X PBS for Tempus 50mL. Box 2 -
  • Stop Solution 2mL
  • DNase 1 200 μL


Requires : Standard centrifuge capable of 5,000 x g with either 96-well plate or microfuge tube adaptors and 96-well plate or microfuge tube vortexer

Description

Description

The Stabilized Blood-to-CT Nucleic Acid Preparation Kit for qPCR makes it possible to perform expression analysis directly from a 500 μL aliquot of human whole blood collected in Tempus Blood RNA tubes without conventional RNA purification. This simple procedure creates a lysate that can go directly into reverse transcription and real-time PCR with the sensitivity and specificity equivalent to conventionally purified RNA. This kit saves time, generates much less waste, and offers a simple workflow that is suitable for a few samples or can easily be incorporated into automated, high throughput applications.

• Detect total RNA (including miRNA) from Tempus™ Blood RNA Tubes without purification
• Robust—500μL sample processed at room temperature with results equivalent to those from purified RNA
• Flexible Format—Samples can be prepared in microfuge tubes or manual/automated 96-well plates

Simple Workflow Without Sacrificing Sensitivity & Specificity
The process consists of pelleting and washing a 500uL aliquot from the blood tube, followed by a unique digestion/DNase step which preserves RNA integrity. The preparation takes place entirely at room temperature unlike other products that require temperature-specific incubation steps. Up to 96 samples can be prepared in half the time it takes for competitive spin column purification to process only 12 samples, with less hands-on effort. The resulting lysate can be stored for later use (1 hour at 4°C or 5 months frozen). This straightforward procedure takes place in the same sample processing well, from start to finish, meaning no sample transfers. The real-time RT-PCR data generated from the lysate is equivalent to results obtained from conventionally purified RNA from the same tubes and has been validated with the following qPCR reagents:
• Applied Biosystems™ High Capacity RNA-to-cDNA™ Kit and TaqMan™ Gene Expression Master Mix
• Invitrogen's SuperScript™ VILO™ cDNA Synthesis Kit and TaqMan Gene Expression Master Mix
• Applied Biosystems™ TaqMan MicroRNA Reverse Transcription Kit and TaqMan Universal Master Mix II
• Applied Biosystems™ TaqMan™ One-Step RT-PCR Master Mix Reagents Kit
Reverse transcription reagents for cDNA synthesis, TaqMan master mixes, and TaqMan assays for real-time PCR analysis are sold separately and are not included with the kit.

The total number of reverse transcription and PCR reactions possible from one starting sample depends on the minimum sample requirements of the desired RT-PCR kits (Refer to user documentation for details).

Screen, Discover, or Validate
For users with specific genes of interest, the Stabilized Blood-to-CT™ Nucleic Acid Preparation Kit for qPCR was designed to enable quick screening from a small amount of starting sample (approximately 5% of the total volume of a properly filled Tempus Blood RNA Tube per sample). The remainder of the blood tube can be archived for later use or purified as needed.

Those who perform discovery or validation experiments with real-time RT-PCR analysis will save effort, time, and overall cost with the Stabilized Blood-to-CT Nucleic Acid Preparation Kit for qPCR – while employing a scalable process allowing labs bottlenecked by conventional purification to finally start processing samples with ease.

miRNA Analysis
With the growing focus on non-coding RNA and its regulatory role in development and disease, it's reassuring to know these small but important RNA molecules can be detected properly with the Stabilized Blood-to-CT Nucleic Acid Preparation Kit for qPCR. Our scientists have tested the lysate with the Applied Biosystems TaqMan MicroRNA Reverse Transcription Kit and TaqMan Universal Master Mix II against a large panel of TaqMan™ microRNA assays and generated results equal to or better than those of the leading competitor's miRNA purification product.

Reduced Footprint & Less Waste = More Savings
The Stabilized Blood-to-CT Nucleic Acid Preparation Kit for qPCR simplifies workflow and reduces total plastic waste by 75% (by weight) for a comparable number of samples processed via columns and generates no organic waste. Space-saving footprint maximizes freezer capacity.

For Users Needing Full Purification
If conventional RNA purification is still required for the remnant blood tube, use the MagMAX for Stabilized Blood Tubes RNA Isolation Kit. This kit maximizes yield of total RNA from Tempus Blood RNA Tubes using magnetic bead-based separation technology.

Blood tubes, reverse transcription reagents for cDNA synthesis, and TaqMan™ master mixes for real-time PCR analysis sold separately.

Specifications

Specifications

Spin Column
Kit
Lysate
Fewer resources used and less waste, Less hazardous
200 Preps
Ambion™, Blood-to-CT™
200 Preps
< 1 hr.

Kit Components:
Box 1-Store at 4°C
• Digestion Solution, 20 mL
• Tempus™ Pellet Enhancer, 4 mL
• Tempus™ Wash #1, 150 mL
• Tempus™ Wash #2, 150 mL
• 2X PBS for Tempus™, 50 mL

Box 2-Store at -20°C
• Stop Solution, 2 mL
• DNase 1, 200 μL

Blood
Real-Time Quantitative PCR (qPCR)
High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
1-step qRT-PCR, 2-step qRT-PCR
Nucleic Acid Preparation Kit
Lysate, ready for real-time PCR
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.