missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Ambion™ T4 Polynucleotide Kinase (cloned) 10U/μL
Catalyzes the transfer of the terminal phosphate of ATP to 5' hydroxyl termini of DNA or RNA
Brand: Invitrogen™ AM2310
Includes: T4 Polynucleotide Kinase
Description
- T4 PNK (PNK, E.C. 2.7.1.78) is recommended for 5' end-labeling of oligonucleotides and polynucleotides
- It also has a 3' phosphatase activity. One tube containing 500U (10U/μL) is provided. T4 Polynucleotide Kinase is rigorously tested for contaminating nonspecific endonuclease, exonuclease, RNase and protease activity, and for functionality in a KinaseMax Kit (Mfr. No. AM1520)
- Reaction buffer is not included with this enzyme
- Unit Definition: One unit is defined as the amount of enzyme catalyzing the production of 1nmol of phosphate to the 5'-OH end of an oligonucleotide from [γ-32P]ATP in 30 minutes at 37°C
Cloning, DNA & RNA Purification & Analysis, Nucleic Acid Labeling & Oligo Synthesis, Oligo Labeling, Restriction Enzyme Cloning
Order Info
Shipping Condition: Dry ice
For Research Use Only. Not for use in diagnostic procedures.