missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ T7 RNA Polymerase, cloned, 200 U/μL

Product Code. 10026284
Click to view available options
Quantity:
30,000 units
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10026284

Brand: Invitrogen™ AM2085

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

This cloned high-purity Ambion™ T7 RNA Polymerase has a high specificity for its promoter.

The enzyme will transcribe large amounts of RNA from DNA sequences downstream of the promoter and show no cross talk between promoters. One tube containing 30,000 U at a concentration of 200 U/ μL is provided. Its high purity makes them ideal for synthesizing high specific activity RNA hybridization probes, biologically active mRNA, and antisense RNA. The RNA Polymerase is rigorously tested for contaminating nonspecific endonuclease, exonuclease, protease, and RNase activity. Functionality of enzyme is tested in In Vitro Transcription reaction under optimal and limiting UTP concentration.

Unit Definition:
One unit is the amount of T7 RNA polymerase required to catalyze the incorporation of 1 nmol of nucleoside triphosphate in 60 minutes at 37°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C.
Shipping Condition Dry Ice
Polymerase T7 RNA Polymerase
Product Line Ambion
Quantity 30,000 units

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.