Learn More
Invitrogen™ T7 RNA Polymerase-Plus™ Enzyme Mix (200 U/μL)
High-purity enzyme that's a mix of cloned RNA polymerase and RNase inhibitor
Brand: Invitrogen™ AM2718
430.65 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 50
Additional Details : Weight : 0.10500kg
Includes: One tube containing 30,000 units at a concentration of 20U/μL
Description
- This enzyme has a high specificity for its promoter, transcribing large amounts of RNA with no cross talk between promoters
- It is supplied in one tube containing 30,000U at a concentration of 200U/μL
- The enzyme incorporates radiolabeled nucleotides along with many modified nucleotides
- Inclusion of RNase inhibitor provides broad-spectrum protection against enzymatic degradation of the synthesized RNA, making the enzyme ideal for the synthesis of RNA for blot and in situ hybridization probes, RPA analysis, in vitro translation, and antisense RNA
Unit Definition: One unit is the amount of T7 RNA polymeras T7 RNA Polymerase-Plus™ Enzyme Mix required to catalyze the incorporation of 1 nmol of nucleoside triphosphate in 60 minutes at 37°C.
DNA & RNA Purification & Analysis, Gene Expression Analysis & Genotyping, in vitro Transcription, Nucleic Acid Labeling & Oligo Synthesis, RNA Labeling
Order Info
Shipping Condition: Dry ice
Specifications
Store at –20°C. | |
T7 RNA Polymerase | |
T7 | |
200 U/μL |
Dry Ice | |
Ambion™ | |
200 μL | |
DNA |
For Research Use Only. Not for use in diagnostic procedures.