Learn More
Invitrogen™ TaqMan™ PreAmp Cells-to-CT™ Kit
The TaqMan PreAmp Cells-to-CT Kit makes it possible to perform gene expression analysis directly from limited or small numbers of cultured cells without RNA purification.
Brand: Invitrogen™ 4387299
Code : 50
Additional Details : Weight : 0.13000kg
Includes: 40 sample prep rxns, 40 Dnase rxns, 40 reverse transcription rxns, and 100 20μL PCR rxns for 40 reactions.
Description
- Complete, validated solution: Optimized workflow includes cell lysis reagents, DNase, RT reagents, PreAmp reagents, and TAqMan Gene Expression Master Mix
- Fast, simple and convenient: Prepare samples in 7 minutes; eliminate tedious RNA isolation
- Extend limited samples: Increase real-time PCR potential by up to 64-fold; study up to 100 genes at once
- Accurate results: Uniform amplification without introducing bias
- Robust performance: Linear detection from 10 to 100,000 cells; results equivalent to purified RNA
- Fewer resources used, less hazardous, less waste
Order Info
Shipping Condition: Dry ice
Specifications
• 40 x 20 μL sample prep reactions |
|
Kit | |
2-step RT-qPCR | |
Standard | |
Fewer resources used and less waste, Less hazardous | |
Ambion™, Cells-to-CT™, TaqMan™ | |
UP (Ultra Pure) | |
Dry Ice | |
Pre-amplification | |
Cell |
Primer-probe | |
Low | |
DNA Polymerase | |
7000 System, 7300 System, 7500 System, 7900HT System,Applied Biosystems StepOnePlus™ Fast Real-Time PCR System, StepOne™, Standard Mode, StepOnePlus™, Standard Mode, QuantStudio 6 and 7 Pro systems, QuantStudio 7 Flex system, QuantStudio 3 and 5 systems, QuantStudio 12k Flex system, QuantStudio 6 Flex system | |
ROX (Pre-mixed) | |
Preamp Cells-to-C Kit | |
M-MLV | |
40 Reactions | |
40 Reactions |
For Research Use Only. Not for use in diagnostic procedures.