missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ THE RNA Storage Solution

Product Code. p-4926938
Click to view available options
Quantity:
50 mL
10 x 1 mL
Unit Size:
10mL
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10449224

Brand: Invitrogen™ AM7000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The Invitrogen RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer.

The Invitrogen RNA Storage Solution is a buffer that provides greater RNA stability than standard 0.1 mM EDTA or TE Buffer. This 1 mM sodium citrate, pH 6.4 ± 0.2 buffer is provided in one bottle containing 50 mL.

The last step in every RNA isolation protocol is to resuspend the purified RNA pellet. After painstakingly isolating the RNA, it is crucial that the pellet be suspended and stored in a safe, RNase-free solution. The RNA Storage Solution has two features that minimize base hydrolysis of RNA: a low pH, and sodium citrate, which is an efficient chelating agent. The RNA Storage Solution is compatible with all common RNA applications such as reverse transcription, in vitro transcription, northern analysis, and nuclease protection assays. The RNA Storage Solution is prepared from the purest reagents available, under scrupulously monitored conditions in a state-of-the-art manufacturing facility. It is then subjected to rigorous quality control procedures to guarantee their purity and performance.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1 mM
Content And Storage Store at -20°C.
Shipped at ambient temperature.
Packaging Type Tube
Product Line Invitrogen
Product Type RNA Storage Reagent
Quantity 10 x 1 mL
Shipping Condition Room Temperature
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.