missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Thin-walled, frosted lid, RNase-free PCR tubes (0.2 mL)

Thin-walled, flat, frosted cap provides a suitable surface for detailed labeling.

Brand:  Invitrogen™ AM12225

140.70 GBP valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10085764

  • £201.00 / Pack of 1000

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Thin-walled, flat, frosted cap provides a suitable surface for detailed labeling. 0.2 mL tubes are provided in a box of 1000. Each lot of tubes is certified RNAse- and DNase-free.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear
1000 tubes
Ambion™
Store at room temperature.
Fast
Flat Cap
Tube
0.2 mL
Thermal Cyclers
Polypropylene
RNase-Free
Room Temperature
Yes
RNase-free PCR Tube
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.