Learn More
Invitrogen™ Tris (1 M), pH 8.0, RNase-free
Invitrogen Molecular biology grade, 1M Tris, pH 8.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use.
Brand: Invitrogen™ AM9856
74.67 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Invitrogen Molecular biology grade, 1M Tris, pH 8.0 solution is supplied in one bottle. The buffer is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.
Order Info
Shipping Condition: Room temperature

Specifications
1 M | |
Liquid | |
500 mL | |
Molecular Biology |
8 | |
Ambion | |
1,3-Propanediol, 2-Amino-2-(Hydroxymethyl)-,Hydrochloride |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Special Offers
-
Multiple Manufacturers
Chemical Extravaganza
Shop now and save up to 97% on top-quality products for your lab needs.Promo Code: 25908Expires: 2025-03-31
-
Multiple Manufacturers
New Offer Save Now
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.