missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TURBO DNase™ (2 U/μL)

Product Code. 10646175
Click to view available options
Quantity:
1000 U
5000 U
Unit Size:
5000 units
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10646175

Brand: Invitrogen™ AM2238

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Efficiently degrades and digests DNA

Cleaves double-stranded DNA nonspecifically to leave 5' phosphorylated oligodeoxynucleotides with an increased affinity for DNA-binding and remains active in presence of salt.

  • Up to 50X more activity and 350% greater catalytic efficiency
  • Efficiently degrades DNA in solutions containing up to 0.25M salt
  • Efficiently digests DNA to oligonucleotides
  • Used in clearing DNA templates from in vitro transcription reactions
  • RNase-free and recombinant in origin
  • Used to clear DNA contamination from RNA samples prior to RT-PCR
  • Conventional DNase I has a poor affinity for DNA and cleaves DNA of low concentration
  • Salt-sensitive; 20 mM NaCl can reduce activity of enzyme by 30%
  • Purified from bovine pancreas, one of richest natural sources of RNase A
  • Threat of contaminating RNase activity in DNase I preparations requires that enzyme be exhaustively purified

TURBO DNA-free (Mfr. No. AM1907) is available. It contains TURBO DNase along with reagents to inactivate the enzyme and remove divalent cations from the sample post-digestion.

PCR and Real-Time PCR, Real Time PCR (qPCR), Reverse Transcription

Order Info

Shipping Conditions: Dry ice

TRUSTED_SUSTAINABILITY

Specifications

Concentration 2 U/μL
Content And Storage TURBO™ DNase and 10X Reaction Buffer should be stored at –20°C.
Shipping Condition Dry Ice
Enzyme DNase
Compatible Buffer 10X Reaction Buffer
Quantity 1000 U
Product Type DNase
Product Line Ambion, TURBO

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.