missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Yeast tRNA (10 mg/mL)

Product Code. 10618255
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10618255

Brand: Invitrogen™ AM7119

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Invitrogen Yeast tRNA purified from brewer's yeast. When used at a final concentration of 10 to 20 μg/mL, yeast tRNA is an effective coprecipitant to aid in recovery of small amounts of nucleic acids.

Invitrogen Yeast tRNA purified from brewer's yeast. When used at a final concentration of 10 to 20 μg/mL, yeast tRNA is an effective coprecipitant to aid in recovery of small amounts of nucleic acids. It is supplied in one tube containing 0.5 mL at a concentration of 10 mg/mL.

What is a Coprecipitant?
Coprecipitants are inert substances used to aid recovery of nucleic acids during alcohol precipitations. While they can be used for precipitating large amounts of nucleic acids, they are essential for quantitative recovery of small amounts of nucleic acids in dilute solutions. Often, the use of such molecules is desirable for no other reason but visualization of the pelleted precipitate after centrifugation.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C.
Concentration 10 mg/mL
For Use With (Application) Carrier for Nucleic Acid Precipitation
Product Type Yeast tRNA
Product Line Ambion
Quantity 0.5 mL
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.