missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ UltraPure™ BSA (50 mg/mL)

Product Code. 10536735
Click to view available options
Quantity:
1 x 50 mg
5 x 50 mg
Unit Size:
250mg
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10536735

Brand: Invitrogen™ AM2618

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.

Ambion Ultrapure BSA is a 'non-acetylated' BSA pure enough to use when DNA or RNA integrity is critical.

• Tubes containing 50 mg are provided, at a concentration of 50 mg/mL
• Bovine Serum Albumin (BSA) has many uses as a carrier protein and as a stabilizing agent in enzymatic reactions
• In northern, Southern, and dot blot hybridizations, BSA is also used as a blocking agent
• It is recommended in buffers for nick translation, polymerase reactions, and ligations

BSA is also a common additive for PCR amplifications, footprinting, and gel shift assays. In restriction digests, BSA has been shown to enhance enzyme activity. The molecular weight is 68 kD. Ultrapure BSA is rigorously tested for DNase, RNase, endonuclease, protease, peroxidase, and alkaline phosphatase activity, and assayed for fluorescence background. Exonuclease Activity: 3' - 5'.

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at –20°C.
Concentration 50 mg/mL
Product Line Ambion
Quantity 5 x 50 mg
Shipping Condition Dry Ice
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.