missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Wash Solution II, RNA Purification

Product Code. 15554314
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
This item is not returnable. View return policy

Product Code. 15554314

Brand: Invitrogen™ 4305890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TransPrep Chemistry replacement solution

Applied Biosystems replacement solution for use with TransPrep Chemistry is for the isolation on the ABI PRISM™ 6100 Nucleic Acid PrepSation.

DNA and RNA Purification and Analysis, General RNA Purification Reagents and Accessories, RNA Extraction, Total RNA Isolation, Total RNA from Animal Cells and Tissues, Total RNA from Blood

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage

Store at 4°C.

1,000 mL is provided.

Buffer Wash Buffer
Quantity 1000 mL
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.