missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMDEX Goat Anti-Rabbit IgG - Horseradish Peroxidase, 0.5ml Secondary Antibody, Cytiva
Brand: Life Science products Cytiva RPN4301
1018.56 GBP valid until 2024-12-31
Use promo code "21890" to get your promotional price.
Code : GE
Additional Details : Weight : 0.50000kg
Description
Host Species: Goat
Isotype: IgG
Species Reactivity: Rb
Enzyme Immunoassay
Specifications
IgG | |
HRP | |
For ELISA, it is recommended that the conjugate should be diluted in the range 1:1000 to 1:4000 | |
Affinity Purified | |
Secondary |
Monoclonal | |
Goat Anti-Rabbit IgG - Horseradish Peroxidase | |
Goat | |
0.5 mL | |
IgG |
Safety and Handling
Product Identifier
Hazard Statement
Supplemental information
- Goat Anti-Rabbit IgG - Horseradish Peroxidase, 0. 5 ml
Hazard Statement
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN ACUTE-53.9 per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.