Learn More
AMDEX sheep anti-mouse IgG-HRP Secondary Antibody, Cytiva
Brand: Life Science products Cytiva RPN4201
Code : GE
Additional Details : CAS Number : 1185-53-1 Weight : 0.50000kg
Description
Host Species: Sheep
Isotype: IgG
Species Reactivity: Ms
Enzyme Immunoassay
Specifications
IgG | |
HRP | |
For ELISA, it is recommended that the conjugate should be diluted in the range 1:4000 to 1:8000 | |
Affinity Purified | |
Secondary |
Monoclonal | |
Sheep Anti-Mouse IgG - Horseradish Peroxidase | |
Sheep | |
0.5 mL | |
IgG |