missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amdex Streptavidin-HRP Enzyme Conjugate
For ELISA signal amplification. The conjugates are based on novel chemistry which facilitates conjugation of multiple enzyme molecules (alkaline phosphatase or horseradish peroxidase) to a single streptavidin or antibody molecule via a dextran backbone.
Brand: Cytiva RPN4401
978.85 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
This item is not returnable.
View return policy
Description
The conjugates are based on novel chemistry which facilitates conjugation of multiple enzyme molecules (alkaline phosphatase or horseradish peroxidase) to a single streptavidin or antibody molecule via a dextran backbone.
- 10- to 100-fold increase in signal intensity
- Compatible with traditional ELISA systems
- Option to reduce incubation times
- Well controlled nonspecific binding properties
Specifications
HRP | |
500 μL | |
Liquid | |
Stabilized in 50% TRIS Buffer and 50% HRP Conjugate Stabilizer |
0.5 mL | |
2°C to 8°C | |
For use in ELISA systems | |
Strepavidin HRP |