missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ AlkPhos Direct™ Labeling Modules

Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes. AlkPhos Direct Labeling Systems are based on the rapid, direct labeling of DNA or RNA probes with thermostable alkaline phosphatase.

Brand:  Cytiva RPN3681

Code : GE

Additional Details : Weight : 1.20000kg

 View more versions of this product

Product Code. 10074344

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

DNA or RNA probes are labeled during a 30-minute reaction and resulting probes can be used without purification. Hybridization stringency can be controlled by temperature as well as salt concentration.

  • Hybridization stringency can be controlled by elevating temperatures up to 75°C
  • Detection of 60 fg target DNA (single-copy gene) in a genomic Southern blot
  • Rapid protocol saves two to three hours over conventional indirect methods
Specifications

Specifications

2°C to 8°C
Fast and convenient systems for all high-sensitivity applications using DNA, RNA or oligonucleotide probes
Labeling reagent, crosslinker, control DNA, reaction buffer, hybridization buffer and blocking agent
For 5000 cm2 membrane
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers