Learn More
Cytiva Amersham™ CyDye™ Antibody Labeling Kits
Convenient way to tag antibodies with CyDye fluorescent dyes
Brand: Cytiva PA33000
This item is not returnable.
View return policy
Description
CyDye fluorescent dyes are supplied as highly water soluble, reactive NHS esters for easy conjugation to biomolecules containing aliphatic amino groups.
- Conjugations are carried out under mild conditions and are complete within 1 hour
- Dyes are packaged in premeasured amounts and foil sealed to ensure consistent labelings
Order Info
Antibody kits contain all reagents required for 2 x 1mg reactions. Monoclonal antibody kits contain all reagents required for 2 x 0.1mg reactions.
Specifications
For the conjugation of proteins with Cy3 Reactive Dye | |
Cy3 Ab Labeling Kit | |
949.11 | |
2°C to 8°C, protected from light | |
Red |
Each | |
7 to 7.3 | |
Cy3 dye: Soluble in cold water and hot water Coupling buffer: Soluble in cold water and hot water Elution buffer: Soluble in cold water and hot water | |
Cy3 Reactive Dye |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1