Learn More
Cytiva Amersham™ CyDye™ Fluorescent Nucleotides
Label DNA and RNA probes. Cytiva Life Sciences™ Amersham™ CyDye™ Fluorescent Nucleotides are optimized for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.
Brand: Cytiva 25-8010-87
Code : GH
Additional Details : Weight : 0.01200kg
Description
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
- Low nonspecific binding
- High photostability
- High water solubility
- pH insensitive
Specifications
CyDyes are a group of bright fluorescent dyes which can be used in genomic and protein research. Cy dye labelled nucleotides exhibit the following properties: | |
791.99 (Monofunction), 975.15 (Bisfunction) | |
Fluorescent Labeling Reagents | |
-20°C | |
Far-Red |
7.4 | |
Cy5-CTP | |
670nm | |
100 nmol | |
Genomic and Protein research, labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping and gene mapping |