Learn More
Cytiva Amersham™ CyDye™ Fluorescent Nucleotides
Label DNA and RNA probes. Cytiva Life Sciences™ Amersham™ CyDye™ Fluorescent Nucleotides are optimized for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.
Brand: Cytiva PA53031
Code : GH
Additional Details : Weight : 0.02500kg
Description
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
- Low nonspecific binding
- High photostability
- High water solubility
- pH insensitive
Specifications
7 | |
Cy3-dCTP | |
-15°C to -30°C | |
Orange |
765.95 (Monofunction), 949.11 (Bisfunction) | |
570nm | |
250 nmol | |
Genomic and Protein research, labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping and gene mapping |