Learn More
Cytiva Amersham™ CyDye™ Fluorescent Nucleotides
Label DNA and RNA probes. Cytiva Life Sciences™ Amersham™ CyDye™ Fluorescent Nucleotides are optimized for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping.
Brand:  Cytiva PA55032
Code : GH
Additional Details : Weight : 0.02300kg
 View more versions of this product
Product Code. 10723977
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
For labeling DNA and RNA probes especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping, and gene mapping. CyDye labeled nucleotides are available in a range of bright, intense colors with narrow emission bands—ideal for multiplexing within a single sample.
  • Low nonspecific binding
  • High photostability
  • High water solubility
  • pH insensitive
Specifications
Specifications
7
Cy5-dUTP
-15°C to -30°C
Far-Red
791.99 (Monofunction), 975.15 (Bisfunction)
670nm
250 nmol
Genomic and Protein research, labeling DNA and RNA probes, especially for multicolor analysis in microarrays, FISH, chromosome identification, whole chromosome painting, karyotyping and gene mapping