missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs

Product Code. p-3676017 Shop All Cytiva Products
Click to view available options
Label or Dye:
Cy2 bis-Reactive Dye
Cy3 mono-Functional Dye
Cy3.5 mono-Functional Dye
Cy5 Reactive Dye
Cy5 mono-Functional Dye
Cy5.5 bis-Functional Dye
Cy5.5 mono-Functional Dye
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 11798268

Brand: Cytiva PA23501

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Use for the most accurate labeling. Cytiva Amersham™ CyDye™ Reactive Dye 5-Packs contain derivatized CyDye that carries only one reactive group on each dye molecule for accurate labeling of amine groups.

 

    • Convenient way to label antibodies, proteins, drugs, and oligonucleotides with CyDye Fluorescent Dyes.
    • For use in ELISA, immunoprecipitation, and blotting applications.
    • Dyes are packaged in premixed amounts and foil-sealed to ensure consistent labelings.

 

Order Info

Each kit contains five vials of dye. Each vial is sufficient for labeling 1mg protein or 30nmol amino groups.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For the labelling of biological compounds with Cy3.5 monofunctional dye
Quantity Each
Product Type Cy3.5 Mono-Reactive Dye Pack
Kit Contents Five foil packs each containing dried dye to label 1mg of Protein
Sufficient For Labeling 1mg Protein or 30nM Amino groups
Molecular Weight (g/mol) 1102.37
Content And Storage 2°C to 8°C, protected from light
Label or Dye Cy3.5 mono-Functional Dye
Color Red
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.