Learn More
Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)
Suitable for general labeling of amine residues
Brand: Cytiva PA17004
This item is not returnable.
View return policy
Description
Order Info
Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
Specifications
Used for high-throughput screening (HTS) applications | |
Cy7 Bis NHS Ester | |
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. |
10 mg | |
Foil packs, each containing 5, 10 or 50mg dried dye | |
Purple |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Specific target organ toxicity Category 2
Hazard Statement
- H371-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Cy7 (Bis NHS ester)