Learn More
Cytiva Amersham™ CyDye™ Value Packs (bis-Reactive NHS Ester)
Suitable for general labeling of amine residues
Brand: Cytiva PA12000
This item is not returnable.
View return policy
Description
Order Info
Large range of bulk packs available for greatest economy. The packs contain fluors carrying two reactive groups on each dye molecule for general labeling of protein amine groups.
Specifications
Suitable for general labeling of Amine residues | |
Cy2 bis-NHS Ester | |
2°C to 8°C |
5 mg | |
Foil packs, each containing 5, 10 or 50mg dried dye |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1