missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Hydrazide)

For the labeling of free carbonyl groups of glycoproteins and carbohydrates

£308.00 - £1101.00

Specifications

For Use With (Application) For labelling via carbonyl groups. For use in steady state fluorescence, FP and FRET applications
pH Range 3 to 10
Solubility Soluble in cold water and hot water
Content And Storage 2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
View More Specs

Products 5
Product Code Brand Quantity Label or Dye Product Type Price Quantity & Availability  
Product Code Brand Quantity Label or Dye Product Type Price Quantity & Availability  
11585065
View Documents
Cytiva
PA13120
5 mg Cy3 Cy3 Hydrazide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10575735
View Documents
Cytiva
PA13121
1 mg Cy3 Cy3 Hydrazide
£308.00
1mg
In Stock
to see stock.
Add to basket
 
10401095
View Documents
Cytiva
PA15121
1 mg Cy5 Cy5 Hydrazide
£328.00
1mg
In Stock
to see stock.
Add to basket
 
11595065
View Documents
Cytiva
PA15120
5 mg Cy5 Cy5 Hydrazide This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10369524
View Documents
Cytiva
PA15621
1 mg Cy5.5 Cy5.5 Hydrazide
£717.00
1mg
Estimated Shipment: 06-02-2025
to see stock.
Add to basket
 
Description

Description

Hydrazide esters provide a convenient method for the labeling of free carbonyl groups of glycoproteins and carbohydrates. For maximum convenience, stable active hydrazide esters are supplied and stored as solid material.

  • Range of bulk packs available for greatest economy
  • Example protocols included with each dye pack

Specifications

Specifications

For labelling via carbonyl groups. For use in steady state fluorescence, FP and FRET applications
3 to 10
Soluble in cold water and hot water
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles.
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Product Certifications