missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)
For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides
Brand: Cytiva PA15630
View more versions of this product
This item is not returnable.
View return policy
Description
The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.
- Suitable for the labeling of antibodies
- Example protocols included with each dye pack
Antibody labeling
Specifications
For labelling via thiol groups For use in steady state fluorescence, FP and FRET applications | |
Cy5.5 Maleimide | |
3 to 10 | |
Soluble in cold water and hot water | |
Cy5.5 |
25 mg | |
Foil packs, each containing 1, 5 or 25mg dried dye | |
1128.42 | |
2°C to 8°C, protected from light. Reconstituted material may be stored for up to 2 weeks at -20°C in aliquots to avoid repeat freeze thaw cycles. | |
Dark Blue |
Safety and Handling
Signal Word
Hazard Category
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1