missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ CyDye™ Value Packs (mono-Reactive Maleimide)

For the selective labeling of free sulfhydryl groups such as cysteine residues in peptides, proteins, and oligonucleotides

Specifications

Kit Contents Foil packs, each containing 1, 5 or 25mg dried dye
pH Range 3 to 10
View More Specs

Products 8
Product Code Brand Quantity Label or Dye Product Type Price Quantity & Availability  
Product Code Brand Quantity Label or Dye Product Type Price Quantity & Availability  
10479724
View Documents
Cytiva
PA15631
1 mg Cy5.5 Cy5.5 Maleimide N/A
N/A
11515085
View Documents
Cytiva
PA15636
25 mg Cy5.5 Cy5.5 Maleimide N/A
N/A
11545085
View Documents
Cytiva
PA15136
25 mg Cy5 Cy5 Maleimide N/A
N/A
11515095
View Documents
Cytiva
PA13130
5 mg Cy3 Cy3 Maleimide N/A
N/A
11555085
View Documents
Cytiva
PA15130
5 mg Cy5 Cy5 Maleimide N/A
N/A
11525085
View Documents
Cytiva
PA15630
25 mg Cy5.5 Cy5.5 Maleimide N/A
N/A
10637475
View Documents
Cytiva
PA13131
1 mg Cy3 Cy3 Maleimide N/A
N/A
10248624
View Documents
Cytiva
PA15131
1 mg Cy5 Cy5 Maleimide N/A
N/A
Description

Description

The use of mono-reactive maleimides is particularly suited to antibody labeling since the dye molecules are introduced away from the antibody binding site, increasing the chance that functionality of the labeled molecule will be unimpaired.

  • Suitable for the labeling of antibodies
  • Example protocols included with each dye pack

Antibody labeling

Specifications

Specifications

Foil packs, each containing 1, 5 or 25mg dried dye
3 to 10
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1

SDS
Documents

Documents

Product Certifications