Learn More
Cytiva Amersham™ ECL Advance™ Blocking Reagent
Use for Controlling background noise. Cytiva Life Sciences™ Amersham™ ECL Advance™ is Blocking Reagent controls background noise while retaining a high level of signal intensity.
Brand:  Cytiva RPN418
Product Code. 10106424
  • £93.85 / 40g
Explore more special offers
Description
Description
Use for Controlling background noise. Cytiva Life Sciences™ Amersham™ ECL Advance™ is Blocking Reagent controls background noise while retaining a high level of signal intensity.
Fully compatible with both Hybond™ ECL™ (nitrocellulose) and Hybond-P (PVDF) membranes.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Amersham™
2° to 8°C
40g
For blocking Nitrocellulose and PVDF membranes prior to detection with Amersham ECL, Amersham ECL Prime or Amersham ECL Select western blotting detection reagents
Blocking reagent
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Cytiva Amersham™ ECL Advance™ Blocking Reagent > 40g