Learn More
Cytiva Amersham™ ECL™ Blocking Agent
Use for blocking nitrocellulose and PVDF membranes. Cytiva Life Sciences™ Amersham™ ECL™ Blocking Agent blocks membranes to be used with ECL and ECL Plus™ Western blotting detection reagents.
Brand: Cytiva RPN2125
68.00 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GH
Additional Details : Weight : 0.12060kg
Description
- Contains 40g of membrane blocking agent
- Sufficient for at least 20 miniblots
Specifications
Blocking Nitrocellulose and PVDF membranes to be used with ECL™ and ECL™ PlusTM western blotting detection reagents | |
40g ECL™ blocking agent, sufficient for at least 20 miniblots | |
40000 mg |