missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System

Use for fast and easy detection of medium-to-high-target amounts. Cytiva Life Sciences™ Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction

Brand:  Cytiva RPN3000


 View more versions of this product

Product Code. 10269004

  • £940.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: 45-000-922 and 45-000-923: Labeling reagent; Crosslinker; Control DNA; Blocking agent; ECL detection reagents and ECL Gold hybridization buffer
This item is not returnable. View return policy

Description

Description

 

    • Direct probe labeling in a 10 min reaction, 1 h from hybridization to detection with ECL Direct, Hybond N+, and Hyperfilm ECL.
    • Eliminates handling, waste, and regulatory issues associated with the use of radioactivity.
    • No need to strip blots before reprobing.
    • For fast and easy detection of medium- to high-target amounts in applications such as colony/plaque screens, dot blots, and PCR product analyses.
    • Consistent results combining strong signals with very low backgrounds.

 

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2°C to 8°C
DNA labeling reagent 0.5mL; Glutaraldehyde solution 0.5mL; Water 1.0mL; l Hind III control DNA (10μg/mL) 0.1mL; Gold hybridization buffer 500mL; Blocking agent 25g; ECL detection reagent 1 125mL; ECL detection reagent 2 125mL
Labels 5 μg Nucleic Acid
Colony or plaque screens, dot blots and PCR product analyses
ECL Direct Labeling and Detection System
20 min.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
  • H400-Very toxic to aquatic life.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P235-Keep cool.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : glutaral, urea, sodium dodecyl sulphate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.