Learn More
Cytiva Amersham™ ECL™ DualVue Western Blotting Markers
Detect proteins on gel and membrane as well as on film
Brand: Cytiva RPN810
209.00 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : GE
Additional Details : Weight : 0.01400kg
Description
Confirm protein transfer, definite blot orientation and permit calculation of target protein's molecular mass.
- Prestained markers indicate efficiency of protein transfer to membrane and blot orientation
- Tagged recombinant protein markers allow accurate calculation of molecular weight of target proteins
- Compatible with any HRP substrate as well as both PVDF and nitrocellulose membrane types
- Contain a mixture of three prestained colored markers of defined molecular weight (from 15,000 to 100,000) and seven tagged recombinant proteins (molecular weights from 15,000 to 150,000)
Order Info
Each pack contains ECL DualVue markers (125μL) and S-protein-HRP conjugate (50μL) sufficient for 25 standard gel loadings.
Specifications
ECL DualVue Western Blotting Markers contain both colored proteins and tagged proteins that introduce convenience and reliability and facilitate accurate size determination of proteins on Western blots. | |
Liquid | |
Pre-Stained Indicator Proteins, Phosphorylase b: 50μg, 100000 (Mr), Rabbit muscle (source), Myoglobin: 50μg, 16000 (Mr), Horse muscle (source), Lysozyme: 50μg, 15000 (Mr), Chicken Egg white (source) | |
25 Loadings |
Protein Molecular Weight Marker/Ladder | |
Detect proteins on Gel and membrane as well as on film | |
Mr 15 000 | |
-20°C |
Safety and Handling
- EUH210-Safety data sheet available on request.
Supplemental information
- UNKNOWN AQUATIC-Contains 50% of components with unknown hazards to the aquatic environment.