Learn More
Cytiva Amersham™ ECL™ Plex Fluorescent Rainbow Markers
Optimized for use with the ECL Plex Western Blotting Detection System. Amersham™ ECL™ Plex Fluorescent Rainbow Markers are ideal for quantitative western blot.
Brand: Cytiva RPN851E
267.90 GBP valid until 2025-12-31
Use promo code "24372" to get your promotional price.
View more versions of this product
Description
- Monitor progress of protein electrophoresis and assess transfer efficiency and molecular weight of blotted proteins without staining
- Enhanced to enable faster and simpler identification of proteins on SDS-polyacrylamide gels
- Ready-to-load
- Provide sharper, more intense bands on gels and blots and have improved band spacing for more accurate molecular weight determination
- Provides fluorescent images using Cy3 and Cy5 channels
Order Info
Each 120μL pack size is sufficient for use with at least 40 minigels (10 x 8cm) or 20 large gels (20 x 20cm). Each 500μL pack size is sufficient for use with at least 160 minigels (10 x 8cm) or 80 large gels (20 x 20cm).
Specifications
These fluorescent rainbow markers are for use with the ECL Plex Western blotting detection system. | |
Protein Molecular Weight Marker/Ladder | |
For quantitative western blots | |
-20°C |
Multi | |
Supplied in 35% Glycerol and sample buffer containing Mercaptoethanesulphonic Acid (MESNA) as reducing agent | |
500 μL | |
50 Minigel Loadings |
Safety and Handling
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : glicerol, sodium dodecyl sulphate, mensa, glycine N,N'-1,2-ethanediylbis[N-(carboxymethyl)- hydrated
Your input is important to us. Please complete this form to provide feedback related to the content on this product.