missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL™ Prime Western Blotting Detection Reagent
Enables the use of highly diluted primary and secondary antibodies without reducing sensitivity. Cytiva Lifescience™ Amersham™ ECL™ Prime Western Blotting Detection Reagent produces high signal intensity and sensitivity.
£265.00 - £548.00
Specifications
Product Type | ECL Western Blot Detection Reagent |
---|---|
For Use With (Application) | Sensitive Quantitative or Confirmatory Western Blot |
Sensitivity | ∼1 pg |
Content And Storage | 2°C to 8°C |
Includes: Solution A (luminol solution) and Solution B (peroxide solution)
Product Code | Brand | Kit Contents | Sufficient For | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Kit Contents | Sufficient For | Price | Quantity & Availability | ||||
10308449
|
Cytiva
RPN2232 |
Solution A (Luminol solution, 50mL) and Solution B (Peroxide solution, 50mL) | 1000cm2 membrane |
£265.00
Each |
In Stock
Log in to see stock. |
||||
12994780
|
Cytiva
RPN2236 |
Solution A (Luminol solution, 3 × 50mL) and Solution B (Peroxide solution, 3 × 50mL) | 3000cm2 membrane |
£548.00
Pack of 3 |
In Stock
Log in to see stock. |
||||
Description
- Stable signal enables multiple exposures and makes reagent suitable for large experimental series
- Provides convenient handling time between detection and completion of experiment
- Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging), Amersham Hybond™ P (PVDF) membranes and compatible with Amersham Hyperfilm™, Amersham Hybond- ECL (nitrocellulose) membranes
- Also compatible with Rainbow™ Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers
Education and research
Specifications
ECL Western Blot Detection Reagent | |
Sensitive Quantitative or Confirmatory Western Blot | |
∼1 pg | |
2°C to 8°C |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ECL Prime'
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360D-May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : trometamol, ethanediol, imidazole
- UNKNOWN AQUATIC-Contains 4% of components with unknown hazards to the aquatic environment.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title