Learn More
Cytiva Amersham™ ECL Select™ Western Blotting Detection Reagent
Use as a high sensitivity reagent for chemiluminescent Western blotting detection. Cytiva Life Sciences™ Amersham™ ECL Select™ Western Blotting Detection Reagent provides highly intense signal output, for detection of medium to very low protein levels.
Brand: Cytiva RPN2235
282.90 GBP valid until 2024-05-31
Use promo code "21664" to get your promotional price.
Code : GH
Additional Details : Weight : 0.18000kg
Description
Amersham ECL Select is a high sensitivity reagent for chemiluminescent Western blotting detection. High signal intensity makes it suitable for the most demanding Western blotting applications, allowing for detection of minute protein quantities. Signals can be detected with CCD imager or X-ray film. Amersham ECL Select typically exhibits a dynamic range that is complementary to Amersham ECL Prime™, and similar to Amersham ECL Advance™.
- Amersham ECL Select is the most sensitive chemiluminescent Western blotting detection reagent in the Amersham ECL range
- With exceptional signal intensity, Amersham ECL Select results in brighter, clearer bands at low protein levels
- Amersham ECL Select delivers high sensitivity even when using highly diluted primary and secondary antibodies
- Works with a wide range of antibody dilutions, reducing the need for lengthy optimization
- Optimized for imaging with ImageQuant™ LAS 4000 (CCD-based imaging) and compatible with Amersham Hyperfilm
- Compatible with Rainbow Molecular Weight Markers and Amersham ECL DualVue™ Western Blotting Markers
- Optimized for use with Amersham Hybond™ PVDF membranes and compatible with Amersham Protran™ nitrocellulose membranes
Specifications
Chemiluminescent | |
High Signal Intensity Western Blot | |
Solution A (Luminol solution, 50mL), Solution B (Peroxide solution, 50mL) |
Liquid | |
Nitrocellulose, PVDF |
Safety and Handling
- ECL Select
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360D-May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains :
- UNKNOWN ACUTE- per cent of the mixture consists of component(s) of unknown acute toxicity.
- UNKNOWN AQUATIC-Contains % of components with unknown hazards to the aquatic environment.