Learn More
Cytiva Amersham™ ECL™ start Western Blotting Detection Reagent
Reagent is luminol-based chemiluminescent substrate designed for the detection of proteins immobilized to a membrane
Brand: Cytiva RPN3243
110.49 GBP valid until 2024-12-31
Use promo code "21631" to get your promotional price.
Description
Amersham™ ECL™ start is a chemiluminescent detection reagent in the Amersham ECL product family that is recommended for confirmatory analysis and detection of high to medium levels of endogenous proteins. Amersham ECL start has a long signal duration, which enables multiple exposures of the membrane plus a convenient time window between experiment and analysis.
- Long signal duration provides assay design flexibility and robust detection of large experimental sets
- Produces low background, regardless of method of detection
- Working solution is stable for five days
- Long shelf life of 18 hours is convenient for those who run Western blots infrequently
- Detection on X-ray film or CCD Imager
- Recommended Membrane: PVDF or nitrocellulose
- For detection of high abundant proteins
- Colorless detection reagent 1 and 2
Applications:
Routine and confirmatory Western blotting
Specifications
ECL Western Blotting Detection Reagent | |
200 mL | |
Routine and confirmatory Western blotting | |
18°C to 30°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.