missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Amersham™ ECL™ Western Blotting Detection Reagents
Use for all routine Western blotting. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world's first commercially available chemiluminescent detection reagents and still one of the most widely used.
Brand: Cytiva RPN2106
286.11 GBP valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
- Detectable on Hyperfilm™ ECL or with CCD imager
- Uses enhanced luminol-based detection which is suitable for all routine Western Blotting
Western Blotting
Specifications
ECL Western Blot Detection Reagent | |
250 mL | |
Detection of 4000cm2 of membrane | |
2°C to 8°C |
Western Blot | |
∼10 pg | |
250mL detection reagent, 1250mL detection reagent 2 |