Learn More
Cytiva Amersham™ Hypercassette™ Autoradiography Cassette
Use for the toughest demands of research in the life sciences. Cytiva Life Sciences™ Amersham™ Hypercassette™ Autoradiography Cassettes are constructed from strong aluminum and a choice of colors enables color coding of work.
Marke:  Cytiva RPN11648
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 10035854
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen »
Entdecken Sie weitere Sonderangebote
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Beschreibung
Beschreibung
Amersham Hypercassette autoradiography cassettes are constructed from strong aluminum and designed for the toughest demands of life science research. Deep cassettes are 1.5cm in depth, allowing them to accommodate thicker samples. A choice of Hypercassette colors enables color coding of work.
  • Provide maximum contact between film and sample
  • Nonporous pad gives a wipe clean surface, ensuring minimum contamination
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Hypercassette, Neutral (Standard)
5 x 7 in.
Light-tight exposures at temperatures below -70°C
Amersham hypercssette autoradiography cassettes are designed to survive the toughest demands of research in the life sciences.
Neutral (Standard)
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Sonderangebote
Sonderangebote